Precision tubings, Versilon™ C-210-A
Supplier: Saint Gobain Life Sciences
Certificates
About this item
Polyurethane, formulation C-210-A, translucent.
- Consistently tight dimensional tolerance
- Excellent abrasion and tear resistance
- Excellent resistance to oils, greases and fuels
- Retains flexibility in sub-zero environments
- High tear resistance
A strictly controlled manufacturing process ensures this hose has a consistently low dimensional tolerance. This combined with high elasticity means the tubing can be connected to fittings easily and without any problems.
Temperature resistance: −73 to +93 °C
Hardness (Shore durometer A): 82
Density: 1,20
Certifications: Meets FDA 21 CFR, 177.1680 and 177.2600 criteria for food and beverage.