VWR®, Strips of PCR Tubes and Caps
These PCR tube strips and caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
- Certified free from DNase, RNase and human DNA
- Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.
- Catalog No:
- 732-3484
- 732-3485
- 732-0545
- 732-3391
- 732-1506
- 732-3610
- 732-1517
- 731-0367
- 732-1520
- 732-3608
- 732-3499
- 732-1519
- 732-3609
- 211-0381
- 732-0546
- 732-1518
- 732-1521
- 211-0338
- 732-0552
- 732-0553
- 732-0554
- 211-0339
- 732-0550
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging