Order Entry
Norway
ContactUsLinkComponent
Centrifuge tubes, glass, Quickfit®
Centrifuge tubes, glass, Quickfit®
Catalog # 201-8267
Supplier:  SciLabware
undefined
Centrifuge tubes, glass, Quickfit®
Catalog # 201-8267
Supplier:  SciLabware

Specifications

  • Closure type:
    Without cap
  • Ø×L:
    17×115 mm
  • Capacity:
    10 ml
  • Pk:
    10

Specifications

About this item

Borosilicate glass, conical bottom, with Quickfit® socket.

  • Precision-ground Quickfit® socket to accept tapered stopper
  • Each tube is marked with its weight to the nearest 0,1 g to enable tubes of similar weight to be selected from any batch
  • Can be centrifuged at rcf values up to 3000×g

Joint size: 14/15