VWR®, Stoppers for Standard Ground Joints
Greaseless closure, which prevents ground joints from seizing. With inclined, circumferential sealing lips for better sealing and reduced risk of breakage. Easily detachable. With ergonomically shaped grip.
- PTFE, solid
- Excellent chemical resistance
- Gas, liquid and vacuum-tight
- Temperature resistant from –200 to +300 °C