Stoppers for conical ground joints
: KARTELL
Stoppers with coloured insert in the holding disc for better identification.
- Transparent LDPE
- Good chemical resistance
- Sealing rings for impermeable and secure fit
Pentagonal holding disc for easy loosening and removal of the stopper.