Splash heads, Stutzer, straight
Leverandør: Lenz Laborglas GmbH & CO.KG
Sertifikat
Om dette produktet
DURAN®, borosilicate 3.3 glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Place directly onto the distillation flask to intercept droplets, with two standard ground joints.