Chemical resistant gloves, AlphaTec® 02-100
: Ansell
These LLDPE laminated safety gloves have a five-layered design, protecting against a broad range of chemicals, meeting EN Type A chemical resistance standards and ensuring greater workplace safety.
- Wide spectrum resistance
- 100% inspected: Each glove is individually air pressure tested
- Easy to put on and comfortable to wear
- Ideal for use in spill response kits
Provide extreme resistance against a wide range of chemicals, including biological hazards.
Ideal for use in Life Sciences, metal fabrication and chemical industries.
Colour: White
EN 374 (ABC), Cat. III, AQL 0.065
: EN ISO 374-1:2016 Type A (ABCEFGHIMS), EN ISO 374-5 VIRUS; Cat. III, AQL 0.065
- Catalog No:
- 112-1010
- 112-1011
- 112-0944
- 112-0945
- 112-1009
- 112-0946
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN ISO 374-1:2016 Type A (ABCEFGHIMS), EN ISO 374-5:2016
- EN ISO 374-1:2016 Type A (ABCEFGHIMS), EN ISO 374-5:2016
- EN ISO 374-1:2016 Type A (ABCEFGHIMS), EN ISO 374-5:2016
- EN ISO 374-1:2016 Type A (ABCEFGHIMS), EN ISO 374-5:2016
- EN ISO 374-1:2016 Type A (ABCEFGHIMS), EN ISO 374-5:2016
- EN ISO 374-1:2016 Type A (ABCEFGHIMS), EN ISO 374-5:2016