Order Entry
Norway
Orders LinkContactUsLinkComponent
MicroFunnel™ Filter Funnels
MicroFunnel™ Filter Funnels
MicroFunnel™ Filter Funnels
Certificates
MicroFunnel™ Filter Funnels
15 Options Available Below
Product Details & Documents
Save time with disposable units by eliminating the cleaning and sterilisation required with reusable funnels.
  • Disposable
  • Ideal for aqueous solutions
  • Squeeze separation
MicroFunnel™ filter funnels are ideal for testing any aqueous solution for microbial contamination using the principles of Membrane Filter (MF) technique.
The unique squeeze separation makes membrane retrieval easy. Choose from 0,45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications, 0,2 and 0,45 µm Supor® (polyethersulfone) membrane to minimise binding of inhibitory substances, or Metricel® Black (modified polyethersulfone) membrane for better contrast with light colored colonies. Available in 100 and 300 ml capacity. Graduations marked on outside of cylinder for better visibility and easy measurement. Gamma irradiated. Gamma irradiation dose is 15 to 30 kGy.
Applications include quality control analysis in pharmaceutical production, analysing process water for industrial applications, and testing drinking water or wastewater for contamination.
Filtration area: 13,46 cm²
Ø×H: 100 ml: 61×76 mm with cover; 300 ml: 88×91 mm with cover
Ordering information: Filter funnels are individually bagged except where indicated. Other types of disposable filter funnels are available. See MicroFunnel™ ST disposable filter funnels for a convenient and economical choice for sterility testing within isolators. See MicroFunnel™ Plus disposable filter funnels for funnels designed specifically for water system monitoring - sample cup and filter funnel all in one. Sentino™ Microbiology pump - designed specifically for MicroFunnel™ filter funnel. Contact your Avantor sales representative for more information.
Specifications for Product Family
Specifications
  • Catalog No:
  • 516-8960
  • 516-8963
  • 516-8969
  • 516-1228
  • 516-1229
  • 515-0048
  • 516-1233
  • 515-0050
  • 516-8962
  • 516-1234
  • 516-8965
  • 516-1235
  • 516-8968
  • 515-0158
  • 516-0427
  • Effective filtration area:
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • 13,46 cm²
  • Dimensions:
  • 76H×61W mm
  • 76H×61W mm
  • 89H×87W mm
  • 76H×61W mm
  • 76H×61W mm
  • 76H×61W mm
  • 76H×61W mm
  • 76H×61W mm
  • 76H×61W mm
  • 76H×61W mm
  • 76H×61W mm
  • 89H×87W mm
  • 89H×87W mm
  • 89H×87W mm
  • 76H×61W mm
Specifications
Product Family Options
Product Information
  • Capacity
    Material
    Membrane
    Packed
    Pore size
    Availability
    Price
  • 516-8960
    MicroFunnel™ filter funnel, white, sterile
    MicroFunnel™ filter funnel, white, sterile
    100 ml
    PP/PS/PE with PES-membran
    Supor® 200, gridded
    Sterile, individually bagged
    0,2 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 5 350,00
    kr 5 350,00
    1 * 50 Items
       kr 5 350,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-8960
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 516-8963
    MicroFunnel™ filter funnel, white, Ungridded, sterile
    MicroFunnel™ filter funnel, white, Ungridded, sterile
    100 ml
    PP/PS/PE with PES-membran
    Supor® white, plain membrane
    Sterile, individually bagged
    0,2 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 5 300,00
    kr 5 300,00
    1 * 50 Items
       kr 5 300,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-8963
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 516-8969
    MicroFunnel™ filter funnel, white, sterile
    MicroFunnel™ filter funnel, white, sterile
    300 ml
    PP/PS/PE with PES-membran
    Supor®, gridded
    Sterile, individually bagged
    0,2 µm
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - kr 3 275,00
    kr 3 275,00
    1 * 20 Items
       kr 3 275,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-8969
    Effective filtration area
    13,46 cm²
    Dimensions
    89H×87W mm
  • 516-1228
    MicroFunnel™ filter funnel, white, gamma irradiated
    MicroFunnel™ filter funnel, white, gamma irradiated
    100 ml
    PS
    ME25
    Sterile, per 50 bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 4 975,00
    kr 4 975,00
    1 * 50 Items
       kr 4 975,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-1228
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 516-1229
    MicroFunnel™ filter funnel, white, gamma irradiated, gridded
    MicroFunnel™ filter funnel, white, gamma irradiated, gridded
    100 ml
    PS
    ME25
    Sterile, bulk
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 3 575,00
    kr 3 575,00
    1 * 50 Items
       kr 3 575,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-1229
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 515-0048
    MicroFunnel™ filter funnel, white
    MicroFunnel™ filter funnel, white
    100 ml
    PP/PS/PE with CE-membran
    GN-6 Metricel®, gridded
    Sterile, per 50 bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 3 525,00
    kr 3 525,00
    1 * 50 Items
       kr 3 525,00
    Specifications:
    More
    Specifications
    Cat. No.
    515-0048
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 516-1233
    MicroFunnel™ filter funnel, white, gridded, gamma irradiated
    MicroFunnel™ filter funnel, white, gridded, gamma irradiated
    100 ml
    PS
    ME25
    Sterile, bulk
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 200 Items
    1 * 200 Items - kr 17 675,00
    kr 17 675,00
    1 * 200 Items
       kr 17 675,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-1233
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 515-0050
    MicroFunnel™ filter funnel, white, gridded, sterile
    MicroFunnel™ filter funnel, white, gridded, sterile
    100 ml
    PP/PS/PE with CE-membran
    GN-6 Metricel®, gridded
    Sterile, individually bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 200 Items
    1 * 200 Items - kr 17 075,00
    kr 17 075,00
    1 * 200 Items
       kr 17 075,00
    Specifications:
    More
    Specifications
    Cat. No.
    515-0050
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 516-8962
    MicroFunnel™ filter funnel, black, gridded, sterile
    MicroFunnel™ filter funnel, black, gridded, sterile
    100 ml
    PP/PS/PE with CE-membran
    Metricel® Black, gridded
    Sterile, individually bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 4 825,00
    kr 4 825,00
    1 * 50 Items
       kr 4 825,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-8962
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 516-1234
    MicroFunnel™ filter funnel, white, gridded, gamma irradiated
    MicroFunnel™ filter funnel, white, gridded, gamma irradiated
    100 ml
    PS
    ME25
    Sterile, per 50 bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 4 800,00
    kr 4 800,00
    1 * 50 Items
       kr 4 800,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-1234
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 516-8965
    MicroFunnel™ filter funnel, white, sterile (includes petri dish lid)
    MicroFunnel™ filter funnel, white, sterile (includes petri dish lid)
    100 ml
    PP/PS/PE with CE-membran
    GN-6 Metricel®, gridded *
    Sterile, individually bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items
    Specifications:
    More
    Specifications
    Cat. No.
    516-8965
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm
  • 516-1235
    MicroFunnel™ filter funnel, white, gridded, gamma irradiated
    MicroFunnel™ filter funnel, white, gridded, gamma irradiated
    300 ml
    PS
    ME25
    Sterile, per 20 bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - kr 2 875,00
    kr 2 875,00
    1 * 20 Items
       kr 2 875,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-1235
    Effective filtration area
    13,46 cm²
    Dimensions
    89H×87W mm
  • 516-8968
    MicroFunnel™ MET, black, gamma irradiated
    MicroFunnel™ MET, black, gamma irradiated
    300 ml
    PP/PS/PE with CE-membran
    Metricel® Black, gridded membrane
    Sterile, individually bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - kr 3 400,00
    kr 3 400,00
    1 * 20 Items
       kr 3 400,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-8968
    Effective filtration area
    13,46 cm²
    Dimensions
    89H×87W mm
  • 515-0158
    MicroFunnel™ filter funnel, white, sterile
    MicroFunnel™ filter funnel, white, sterile
    300 ml
    PP/PS/PE with PES-membran
    Supor®, gridded
    Sterile, individually bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - kr 3 225,00
    kr 3 225,00
    1 * 20 Items
       kr 3 225,00
    Specifications:
    More
    Specifications
    Cat. No.
    515-0158
    Effective filtration area
    13,46 cm²
    Dimensions
    89H×87W mm
  • 516-0427
    MicroFunnel™ filter funnel, white, sterile
    MicroFunnel™ filter funnel, white, sterile
    100 ml
    PP/PS/PE with CE-membran
    Supor®, gridded
    Sterile, individually bagged
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - kr 4 600,00
    kr 4 600,00
    1 * 50 Items
       kr 4 600,00
    Specifications:
    More
    Specifications
    Cat. No.
    516-0427
    Effective filtration area
    13,46 cm²
    Dimensions
    76H×61W mm