VWR®, Dishes, Evaporating, Shallow
PTFE, with spout.
Excellent chemical resistance
-
Temperature resistant from –200 to +280 °C
Outer diameter with a tolerance of +/- 1mm
- Exceptionally smooth inner walls to prevent contamination
Thick walled, conical shape. Extremely robust.