Wash bottles, wide neck, Unitary™, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
LDPE, translucent, with PP screw cap.
- Wide neck for quick, safe filling, leakproof closure
- Formed from a single piece, outlet tube can no longer become contaminated or leak
- Jet tip can be cut to increase flow rate
- Good chemical resistance, heat resistant up to 80 °C
Specifications
- Catalog No:
- 215-8111
- 215-8115
- 215-8114
- 215-8113
- 215-8112
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)