Wash bottles, narrow neck, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PTFE/FEP, with ETFE spray cap.
- Excellent chemical resistance
- Heat resistant up to +205 °C, autoclavable
- Leakproof
With excellent chemical resistance and low permeability. Ideal for contamination-free trace analysis work. Free of phthalates. Jet tip can be cut to increase flow rate.
Specifications
- Catalog No:
- 215-8124
- 215-8123
- 215-8122
- 215-8121
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement