Jerricans, wide neck, HDPE
Supplier: hünersdorff GmbH
Certificates
About this item
HDPE, translucent.
- Good chemical resistance, good temperature resistance
- Sealable cap
- All jerricans feature a screw thread and stoppers on the narrow side
The extra wide filling opening (88 mm I.D.) makes cleaning the inside much easier, which is particularly important when storing and transporting sensitive substances. The carrying handle and hinged screw cap ensure this product can be transported easily and securely. The design with a 10-litre capacity features an additional mini spout (18 mm I.D.). UV-stabilised. The 22-litre jerricans are stackable.