Biohazard bag holders, Clavies®, SP Bel-Art
To keep biohazard disposal bags open and ready to use.
- Stainless steel wire frame
- Base in orange PP, which serves as a leakage tray
- Autoclavable at 121 °C to maintain sterility
: Delivered unassembled.
To keep biohazard disposal bags open and ready to use.