Order Entry
Norway
Orders LinkContactUsLinkComponent
Flange clamps
Flange clamps
Supplier:   Lenz Laborglas
Flange clamps
Supplier:  Lenz Laborglas
Certificates
Flange clamps
3 Options Available Below
Product Details & Documents
Plastic, black.
  • Easy to assemble and disassemble
  • Tough construction, good temperature resistance
  • Matching gaskets are available as an option
For fast, secure connection of ball or flat flanges. No gasket.
Product Family Options
Product Information
  • For
    Availability
    Price
  • ball and flat flanges KF/PF 15
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 1 425,00
    kr 1 425,00
    1 * 1 Items
       kr 1 425,00
  • Ball and flat flanges KF/PF 25
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 1 825,00
    kr 1 825,00
    1 * 1 Items
       kr 1 825,00
  • Ball and flat flanges KF/PF 50
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr 3 150,00
    kr 3 150,00
    1 * 1 Items
       kr 3 150,00