Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
Norway
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
ContactUsLinkComponent
Home
1,2,4-Triazole
1,2,4-Triazole
1,2,4-Triazole
1,2,4-Triazole
PROOMM0235.03
:
LGC Standards PROMOCHEM
:
1,2,4-Triazole
PROOMM0235.03
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
100 mg
Seq:
2440
:
:
69.07 g/mol
:
12,09735
:
260 °C (1013 hPa)
:
116…119 °C
:
1.394 g/cm³ (25 °C)
:
206-022-9
:
GHS08
:
GHS07
:
288-88-0
:
C₂H₃N₃
:
MFCD00005228