

1,2,4-Triazole ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:288-88-0
:
Formula:C₂H₃N₃
MDL Number:MFCD00005228
:69.07 g/mol
Merck Index:14,9605
Boiling Pt:260 °C
Melting Pt:118…122 °C
Density:1.394 g/cm³ (25 °C)
Flash Pt:158 °C (316 °F)
Storage Temperature:Ambient
EINECS:206-022-9
:GHS08
:GHS07