Pararosaniline hydrochloride ≥90%
Katalog # 36540.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
About this item
:
:323.82 g/mol
Melting Pt:268…270 °C (dec.)
EINECS:209-321-2
:GHS08
Storage Temperature:Ambient
CAS Number:569-61-9
Formula:C₁₉H₁₈ClN₃
MDL Number:MFCD00001657