9-Acetylanthracene 95%
:784-04-3
:
:C₁₆H₁₂O
:MFCD00001259
:220.27 g/mol
:374 °C (1013 hPa)
:72…76 °C
:1,26 g/cm³ (20 °C)
:Ambient
:212-315-2
- Appearance:Yellow to yellow-brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:72 to 76 °C
- GC:≥94 %
- Iron (Fe):≤0.002 %
- Magnesium (Mg):≤0.002 %
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.02 %
- Heavy metals (as Pb):≤0.001 %
- Insoluble matter:≤0.01 %