Order Entry
Norway
ContactUsLinkComponent
VWR® Glass Balls
VWR® Glass Balls
  332124G
undefined
VWR® Glass Balls
  332124G

 

  • Ø:
    2,5 - 3,5 mm
  • Description:
    Glass balls, 2.5 - 3.5 mm
  • Pk:
    500 g

 

 

Chemically resistant glass balls suitable for use as mixing beads, boiling stones, or packing for distillation columns.