Flat Cut Rimmed Bowls, 304 Grade Stainless Steel
Stainless steel bowls available in 304 grade material, ideal for laboratory and other pharmaceutical industry uses.
- Hygienic design; pressed in one piece, with a flat base and flat rim
- Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
- Optional material grade, surface finish, and TSE certification available upon request
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information.