Potassium iodide 0.1 M (0.1 N) ISO 17034, 17025, 9001
Catalog # CMPLZ14591073.1L
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:166 g/mol
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405