Order Entry
Norway
ContactUsLinkComponent
 
Health
Celite® 545, filter aid, Supelco®
Catalog # 20199-U
Supplier:  Merck
CAS Number:  
undefined
Health
Celite® 545, filter aid, Supelco®
Catalog # 20199-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :60.08 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:272-489-0
 :GHS08
CAS Number:68855-54-9
Formula:SiO₂
MDL Number:MFCD00132803