Order Entry
Norway
ContactUsLinkComponent
Coomassie brilliant blue G-250
Coomassie brilliant blue G-250
Catalog # A3480.0025
Supplier:  PanReac AppliChem
CAS Number:  
Coomassie brilliant blue G-250
Catalog # A3480.0025
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :854.04 g/mol
Density:>1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:228-058-4
CAS Number:6104-58-1
Formula:C₄₇H₄₈N₃NaO₇S₂
MDL Number:MFCD00078482