Order Entry
Norway
ContactUsLinkComponent
 
Coomassie brilliant blue G-250, pure, Sigma-Aldrich®
 
undefined
Coomassie brilliant blue G-250, pure, Sigma-Aldrich®
 

 

  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Dark Blue to Very Dark Blue
  • Solubility (Form), 1 mg/ml, H₂O:
    Solution
  • Suitability, Suitable for use as a reagent in protein determinations:
    Suitable
  • Purity (HPLC):
    ≥80%
  • Recommended Retest Period:
    4 years