Order Entry
Norway
ContactUsLinkComponent
 
Sudan III, Millipore®
  1.11747.0025
 :  Merck
 :  
Sudan III, Millipore®
  1.11747.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Melting Point:
    199 °C (decomposition)
  • Molar Mass:
    352.39 g/mol

 

 

 :
 :352.4 g/mol
 :12,09054
 :205.3…206.3 °C
 :M-Clarity™ quality level = MQ200
 :201-638-4
 :85-86-9
 :C₂₂H₁₆N₄O
 :MFCD00003905