Order Entry
Norway
ContactUsLinkComponent
Nuclear Fast Red
Nuclear Fast Red
Catalog # CHRO1A-402
Supplier:  CHROMA
CAS Number:  
Nuclear Fast Red
Catalog # CHRO1A-402
Supplier:  CHROMA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :357.28 g/mol
Merck Index:13,06512
Melting Pt:300 °C
Storage Temperature:Ambient
EINECS:229-088-0
CAS Number:6409-77-4
Formula:C₁₄H₈NNaO₇S
MDL Number:MFCD00078493