Order Entry
Norway
ContactUsLinkComponent
Collagenase Type IV (1 mg/ml)
Collagenase Type IV (1 mg/ml)
Catalog # STMC07909
CAS Number:  
undefined
Collagenase Type IV (1 mg/ml)
Catalog # STMC07909
CAS Number:  
Specifications
  • Fp:
    100 mL
  • Arter:
    Non-human primate
    Human
    Rat
    Mouse
  • Beskrivelse:
    Collagenase type IV (1 mg/ml)
Specifications
About this item
Cell dissociation reagent.
  • Generate human ES cell and iPS cell clumps suitable for passaging
  • Maintain receptor integrity with a collagenase preparation low in tryptic activity
Generate human embryonic stem cells (ES) or human induced pluripotent stem (iPS) cell clumps suitable for passaging with Collagenase Type IV (1 mg/ml). This enzymatic cell dissociation reagent can also be used in the transfer of human ES cells or iPS cells cultured on a layer of feeder cells to feeder-free conditions. Sourced from Clostridium histolyticum, Collagenase Type IV has low tryptic activity and is frequently used for applications where maintenance of receptor integrity is required.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').