Hyaluronidase
Supplier: STEMCELL Technologies
Certificates
About this item
For hydrolysis of hyaluronic acid.
Hydrolize hyaluronic acids in connective tissue with Hyaluronidase. Frequently used with other proteases, such as collagenase, this polysaccharidase cleaves endo-N-acetlyhexosaminic bonds in hyaluronic acid, as well as chondroitin sulfate A and C, to tetrasaccharide residues. Hyaluronidase consists of a single polypeptide chain of 450 amino acid residues with four disulfide bonds and is glycosylated, containing 5% mannose and 2.17% glucosamine (Borders and Raftery).
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
- Species:Non-human primateHumanRatMouse