Order Entry
Norway
ContactUsLinkComponent
Blood bank reagents
Blood bank reagents
Catalog # ORTH721105
CAS Number:  
Blood bank reagents
Catalog # ORTH721105
CAS Number:  
Specifications
  • Pk:
    3 mL
  • Description:
    Anti-JKa BioClone, 1×3 ml
Specifications
About this item
Qualitative tests for analysing irregular blood group antibodies. Analysing and identifying irregular blood group antibodies are important procedures for ensuring optimum follow-up of transfusions, pregnancy and disease. Comparing the reactivity pattern of individual cells with the published ANTIGRAM® antigen profile helps with identifying antibodies.
  • Monoclonal antibodies for analysing various antigens on human erythrocytes