Biphenyl (PCB No. 0) ≥99.0% (by GC), zone refined: number of passes 24
Catalog # TCIAB0224-1SAMPLE
Supplier: TCI
CAS Number:
Specifications
- Pk:1
- Seq:0014
Specifications
Specification Test Results
- Purity/Analysis Method:>99.0% (GC)
Specification Test Results
About this item
:
:154.21 g/mol
Boiling Pt:256 °C
Melting Pt:70 °C
Flash Pt:113 °C
UN:3077
ADR:9,III
CAS Number:92-52-4
Formula:C₁₂H₁₀
MDL Number:MFCD00003054