Order Entry
Norway
ContactUsLinkComponent
EasySep™ Dead Cell Removal (Annexin V) Kit
EasySep™ Dead Cell Removal (Annexin V) Kit
Catalog # STMC17899
undefined
EasySep™ Dead Cell Removal (Annexin V) Kit
Catalog # STMC17899
Specifications
  • Description:
    EasySep™ dead cell removal (Annexin V) kit, 1×10⁹ cells
  • Pk:
    1 KIT
Specifications
About this item
Immunomagnetic depletion of apoptotic (Annexin V+) cells from culture or tissue preparations.
  • Fast and easy-to-use
  • No columns required
  • Compatible across EasySep™, 'The Big Easy', and EasyEights™ platforms
The EasySep™ Dead Cell Removal (Annexin V) kit is designed to deplete apoptotic (Annexin V+) cells from cell culture or tissue preparations by immunomagnetic negative selection. Unwanted cells are labelled with Annexin V, antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are simply poured off into a new tube. Annexin V binds phosphatidylserine on the outer leaflet of the cell membrane during apoptosis.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').