1,2-Distearoyl-sn-glycero-3-phosphocholine 95%
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:816-94-4
:
Formula:C₄₄H₈₈NO₈P
MDL Number:MFCD00036905
:790.16 g/mol
EINECS:212-440-2
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:white to almost white powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Optical rotation:+5.0° to +7.0° (c=2, Chloroform)