Order Entry
Norway
ContactUsLinkComponent
Sclareol
Sclareol
New Product
Sertifikat
Sclareol
New Product

Spesifikasjonstestresultater

  • Appearance:
    white to light yellow powder and or chunks
  • Infrared spectrum:
    Conforms
  • GC:
    ≥94 %
  • Melting point:
    99°C to 103°C
  • Specific optical rotation:
    -23° to -31° (20°C, 589 nm) (1% in pyridine)

Spesifikasjonstestresultater