Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Nitrobenzenesulphonyl chloride
4-Nitrobenzenesulphonyl chloride 97%
4-Nitrobenzenesulphonyl chloride 97%
:
Thermo Fisher Scientific
4-Nitrobenzenesulphonyl chloride 97%
:
Thermo Fisher Scientific
:
98-74-8
:
:
C₆H₄ClNO₄S
:
MFCD00007445
:
221.62 g/mol
:
143…144 °C/1.5 mm
:
76…80 °C
:
Ambient
:
202-697-9
:
3261
:
8,III
:
GHS05
msds_ALFAA18512
Organic Building Blocks and Intermediates