Order Entry
Norway
ContactUsLinkComponent
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
Catalog # MOLEM29710543
Supplier:  Molekula
CAS Number:  
N-Octyl-β-D-glucopyranoside
Catalog # MOLEM29710543
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :292.37 g/mol
Melting Pt:110 °C
EINECS:249-887-8
 :GHS07
Storage Temperature:Freezer
CAS Number:29836-26-8
Formula:C₁₄H₂₈O₆
MDL Number:MFCD00063288