Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
A1010.0025
:
PanReac AppliChem
:
N-Octyl-β-D-glucopyranoside
A1010.0025
:
PanReac AppliChem
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
292.37 g/mol
:
110 °C
:
249-887-8
:
GHS07
:
Freezer
:
29836-26-8
:
C₁₄H₂₈O₆
:
MFCD00063288