Order Entry
Norway
ContactUsLinkComponent
Nitro blue tetrazolium chloride (NBT)
Nitro blue tetrazolium chloride (NBT)
Catalog # MOLE13727374-1G
Supplier:  Molekula
CAS Number:  
Nitro blue tetrazolium chloride (NBT)
Catalog # MOLE13727374-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :817.65 g/mol
Melting Pt:200 °C
EINECS:206-067-4
 :GHS07
Storage Temperature:Refrigerator
CAS Number:298-83-9
Formula:C₄₀H₃₀Cl₂N₁₀O₆
MDL Number:MFCD00012159