



Boron trifluoride - propanol reagent 10 - 20% for esterification (1 ml×10)
Katalog # TCIAX0037-1SET
Supplier: TCI
CAS Number:
Specification Test Results
- Appearance:Colorless to Light yellow to Light red clear liquid
- Concentration (Gravimetric Method):10.0 to 20.0 w/w%
Specification Test Results
About this item
Merck Index:14,01349