Order Entry
Norway
ContactUsLinkComponent
Methylene blue in 3% acetic acid
Methylene blue in 3% acetic acid
Catalog # STMC07060
CAS Number:  
undefined
Methylene blue in 3% acetic acid
Catalog # STMC07060
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :319.86 g/mol
CAS Number:61-73-4
 :MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008