Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
Water
Water nuclease-free
Water nuclease-free
Water nuclease-free
Leverandør:
STEMCELL Technologies
Sertifikat
Expand image
Water nuclease-free
Leverandør:
STEMCELL Technologies
Om dette produktet
CAS-nummer:
7732-18-5
:
Formel:
H₂O
MDL-nummer:
MFCD00011332
:
18.02 g/mol
Lagringstemperatur:
Ambient