Anti-Complement Factor B Mouse Monoclonal Antibody [clone: 19D118]
Catalog # USBI508233
Supplier: US Biological
New Product
Specifications
- Antibody type:Primary
- Antigen name:Complement Factor B
- Clonality:Monoclonal
- Clone:19D118
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Supplied as a liquid in PBS, pH 7,3, 0,02% sodium azide, 50% glycerol
- Purity:Purified by protein A/G affinity chromatography.
- Storage temperature:May be stored at 4 °C for short-term only. Aliquot to avoid repeated freezing and thawing. Store at −20 °C. Aliquots are stable for 12 months after receipt.
- Shipping temperature:20
- Immunogen:Complement factor B
- Pk:100 µG
Specifications
About this item
Recommended dilution:
Western blot: 1:500 to 1:5000
Immunohistochemistry: 1:20 to 1:200
Optimal dilutions to be determined by the researcher
Specificity: Recognises complement factor B
Species crossreactivity: human
Type: Primary
Antigen: Complement Factor B
Clonality: Monoclonal
Clone: 19D118
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human