Colour Standards, Saybolt
Supplier: LGC Standards PROMOCHEM
New Product
Certificates
About this item
This product is intended for the calibration and/or verification of equipment used for the measurement of colour of liquid samples. This product may also be used for visual based methodology.
- Single solution
- Store at room temperature
- Laboratory use
Certifications: ISO 17025, ISO 17034 Certified.