Order Entry
Norway
ContactUsLinkComponent
Whole Normal Kidney Tissue Lysate (Adult Normal)
Whole Normal Kidney Tissue Lysate (Adult Normal)
Catalog # NOVUNB820-59231
Supplier:  Novus Biologicals
New Product
CAS Number:  
Whole Normal Kidney Tissue Lysate (Adult Normal)
Catalog # NOVUNB820-59231
Supplier:  Novus Biologicals
New Product
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    1 mg
  • Tested applications:
    WB, ImmunoPrecipitation

Specifications

About this item

Whole normal kidney tissue lysate (adult normal).

  • Source: Human

Tested applications: Western blot, Immunoprecipitation. Tested reactivity: Human.