Supelco® Cerilliant® Organic Reference Standard, (-)-Riboflavin-¹³C₄,¹⁵N₂ (Vitamin B2-¹³C₄,¹⁵N₂) solution
Catalog # V-055-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml (1% ammonium acetate in 50:50 methanol:water)
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:1217461-14-7
- Description:(-)-Riboflavin-¹³C₄,¹⁵N₂ (Vitamin B2-¹³C₄,¹⁵N₂) solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- MDL No.:MFCD00005022
- M-Clarity quality level:MQ300
- Molecular weight:382,32
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311