Order Entry
Norway
ContactUsLinkComponent
Anti-CXCR4 Mouse Monoclonal Antibody [Clone: 12G5] (PE (Phycoerythrin))
Anti-CXCR4 Mouse Monoclonal Antibody [Clone: 12G5] (PE (Phycoerythrin))
Catalog # ANTIA86616-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CXCR4 Mouse Monoclonal Antibody [Clone: 12G5] (PE (Phycoerythrin))
Catalog # ANTIA86616-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C-X-C chemokine receptor type 4
  • Antigen symbol:
    CXCR4
  • Clonality:
    Monoclonal
  • Clone:
    12G5
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P61073
  • Antigen synonyms:
    CXCR4_HUMAN|Chemokine (C X C motif) receptor 4|CXCR-4|CXCR4|CXC-R4|D2S201E|CD184 antigen|Chemokine CXC Motif Receptor 4|CD184
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    CP-MAC-infected Sup-T1 cells
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [12G5] antibody to CXCR4 (PE) for Flow Cytometry with samples derived from Human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Non-Human Primates