Anti-Cyclin D1 Mouse Monoclonal Antibody [Clone: CD1.1]
Catalog # ANTIA86601-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:AI327039
- Antigen symbol:Cyclin D1
- Clonality:Monoclonal
- Clone:CD1.1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P24385
- Antigen synonyms:BCL1|B cell leukemia 1|BCL-1 oncogene|B-cell lymphoma 1 protein|B cell CLL/lymphoma 1|B cell lymphoma 1 protein|BCL1 oncogene|BCL-1|BCL 1
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified cyclin D1 protein
- Tested applications:IHC-P, IHC-Fr, ICC
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [CD1.1] antibody to Cyclin D1 for Flow Cytometry, IP, WB, IHC-P, IHC-Fr, ICC and ELISA with samples derived from Human and Rat.
- Validated applications: Flow cytometry, IP, WB, IHC-P, IHC-Fr, ICC, ELISA
- Recommended dilutions: IHC-Fr: 2 µg/ml, WB: 1 µg/ml, ICC: 1 µg/ml, IP: 1 µg/ml
Type: Primary
Antigen: Cyclin D1
Clonality: Monoclonal
Clone: CD1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat