Anti-Crebbp Rabbit Polyclonal Antibody
Catalog # ANTIA93361-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CBP
- Antigen symbol:Crebbp
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q6GQV9
- Antigen synonyms:CREB binding protein|RTS|RSTS|CBP_HUMAN|KAT3A / CBP|Crebbp|Cyclic AMP responsive enhancer binding protein|KAT3A|CREB-binding protein
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Molecular weight:290 kDa
- Sequence:MAENLLDGPPNPKRAKLSSPGFSANDNTDFGSLFDLENDLPDELIPNGELSLLNSGNLVPDAASKHKQLSELLRGGSGSSINPGIGNVSASSPVQQGLGGQAQGQPNSTNMASLGAMGKSPLNQGDSSTPNLPKQAASTSGPTPPASQALNPQAQKQVGLVTSSPATSQTGPGICMNANFNQTHPGLLNSNSGHSLMNQAQQGQAQVMNGSLGAAGRGRGAGMPYPAPAMQGATSSVLAETLTQVSPQMAGHAGLNTAQAGGMTKMGMTGTTSPFGQPFSQTGGQQMGATGVNPQLASKQSMVNSLPAFPTDIKNTSVTTVPNMSQLQTSVGIVPTQAIATGPTADPEKRKLIQQQLVLLLHAHKCQRREQANGEVRACSLPHCRTMKNVLNHMTHCQAGKACQVAHCASSRQIISHWKNCTRHDCPVCLPLKNASDKRNQQTIL
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-445 of mouse CREBBP (Q6GQV9)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Crebbp for WB with samples derived from Human, Mouse and Rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Crebbp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat