Order Entry
Norway
ContactUsLinkComponent
Anti-Glutamate receptor ionotropic, NMDA 2D Rabbit Polyclonal Antibody
Anti-Glutamate receptor ionotropic, NMDA 2D Rabbit Polyclonal Antibody
Catalog # ANTIA95313-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Glutamate receptor ionotropic, NMDA 2D Rabbit Polyclonal Antibody
Catalog # ANTIA95313-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EB11
  • Antigen symbol:
    Glutamate receptor ionotropic, NMDA 2D
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O15399
  • Antigen synonyms:
    GluN2D|Glutamate receptor ionotropic|Glutamate receptor, ionotropic, N-methyl D-aspartate 2D|Glutamate [NMDA] receptor subunit epsilon 4|Glutamate receptor ionotropic, NMDA 2D|Grin2d|Glutamate [NMDA] receptor subunit epsilon-4|estrogen receptor binding CpG island|glutamate receptor ionotropic NMDA 2D
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    143 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human NMDAepsilon4 (amino acids 644-693)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to NMDAepsilon4 for IHC and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: IHC, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: Glutamate receptor ionotropic, NMDA 2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat