Anti-CFTR Mouse Recombinant Antibody [clone: rCFTR/8047]
ANTIA314783-100
New Product
- Antibody type:Primary
- Antigen name:ABC 35
- Antigen symbol:CFTR
- Clonality:Recombinant
- Clone:rCFTR/8047
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P13569
- Antigen synonyms:ABC35|ABCC7|ATP-binding cassette sub-family C member 7|cAMP-dependent chloride channel|cAMP dependent chloride channel|ATP binding cassette transporter sub family C member 7|ABCC 7|ATP binding cassette sub family C member 7|ATP Binding Cassette Superfamily C Member 7
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:165 - 170 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 550-850) of human CFTR protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rCFTR/8047] antibody to CFTR for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: CFTR
Clonality: Recombinant
Clone: rCFTR/8047
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human