Anti-PDXP Goat Polyclonal Antibody
ANTIA84897-100
New Product
- Antibody type:Primary
- Antigen name:Chronophin
- Antigen symbol:PDXP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PDXP|PLP phosphatase|Pyridoxal (pyridoxine, vitamin B6) phosphatase|CIN|Pyridoxal phosphate phosphatase|PLPP_HUMAN|PLPP|PLP|FLJ 32703
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-QHDLVPHYYVES
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human PDXP (C terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to PDXP for ELISA, WB and IHC with samples derived from human, mouse and rat.
- Validated applications: ELISA, WB, IH
Type: Primary
Antigen: PDXP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat